missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramphenicol USP , Medisca™

Supplier: Medisca Pharmaceutique Inc 020304
| Quantity | 25 g |
|---|
Description
Chloramphenicol USP comes as fine, white to grayish-white or yellowish-white, needle-like crystals or elongated plates.
Chemical Identifiers
| 56-75-7 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 56-75-7 | |
| 25 g | |
| USP |
| Crystals | |
| Beige or Gray/Yellow to White | |
| 97.0 to 103.0% |