missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6717FR Evaporative Cooling Hard Hat Pad with Neck Shade

Catalog No. 19819604
Change view
Click to view available options
Certifications/Compliance:
ASTM F2302
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
19819604 ASTM F2302
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819604 Supplier Ergodyne™ Supplier No. 12657

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6717FR Evaporative Cooling Hard Hat Pad with Neck Shade provides cooling relief and FR protection.

  • Protects neck from harmful UV rays
  • Modacrylic cotton blend FR fabric
  • Fabric meets ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2); classified CAT1 per NFPA 70E
  • Meets ASTM F2302 for flame resistance
  • Soak in water 2 to 5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Conveniently attaches to hard hat suspension headband
  • Reusable

Specifications

Certifications/Compliance ASTM F2302
Color Blue
For Use With (Application) Construction, Maintenance, Landscaping/Grounds, Electrical Workers
Material Modacrylic Cotton Blend