missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6717CT Evaporative Cooling Hard Hat Neck Shade with Cooling Towel

Catalog No. 19819650
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819650 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819650 Supplier Ergodyne™ Supplier No. 12596

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6717CT Evaporative Cooling Hard Hat Neck Shade with Cooling Towel provides protection against UV.

  • Advanced PVA material activates easily and quickly when run under water for one minute
  • Hook and loop closures connect to hard hat suspension system
  • Remains hydrated for up to 4 hours
  • Protects neck from harmful UV rays
  • Conveniently attaches to hard hat suspension headband
  • Reusable

Specifications

Color Blue
For Use With (Application) Construction, Landscaping
Material PVA