missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6717 Evaporative Cooling Hard Hat Pad with Neck Shade

Catalog No. 19819501
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819501 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819501 Supplier Ergodyne™ Supplier No. 12336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6717 Evaporative Cooling Hard Hat Pad features a neck shade, which helps protect against the sun.

  • Lightweight, cooling acrylic polymers
  • Hook and loop closures
  • Protects neck from harmful UV rays
  • Conveniently attaches to hard hat suspension system
  • Soak in water 2 to 5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Reactivate by re-soaking

Specifications

Color Blue
For Use With (Application) Construction, Maintenance, Landscaping, Warehousing/Distribution