missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6710FR Evaporative FR Cooling Triangle Hat

Catalog No. 19808125
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19808125 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19808125 Supplier Ergodyne™ Supplier No. 12627

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activated polymers bring cooling relief along with FR protection.

  • Modacrylic cotton blend fabric
  • Fabric meets ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2)
  • Classified CAT1 per NFPA 70E
  • Meets ASTM F2302 for flame resistance
  • Soak in water 2-5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Re-usable, just re-wet
  • Colors: Solid Blue, Hi-Vis Lime

Specifications

Material Modacrylic Cotton Blend FR Fabric
Product Type High Performance Dew Rag
Size Universal
Color Blue
For Use With (Application) Cooling