missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6710FR Evaporative FR Cooling Triangle Hat

Catalog No. 19819596
Change view
Click to view available options
Color:
Lime
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819596 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819596 Supplier Ergodyne™ Supplier No. 12621

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6710FR Evaporative FR Cooling Triangle Hat features activated polymers that bring cooling relief and FR protection.

  • Modacrylic cotton blend fabric
  • Fabric meets ASTM F1506 performance requirements (ATPV rating of 7.1 cal/cm2); classified CAT1 per NFPA 70E.
  • Meets ASTM F2302 for flame resistance
  • Soak in water 2 to 5 minutes to activate
  • Remains hydrated for up to 4 hours
  • Reusable

Specifications

Color Lime
For Use With (Application) Construction, Maintenance, Landscaping, Warehousing/Distribution