missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6710 Evaporative Cooling Triangle Hat

Catalog No. 19819507
Change view
Click to view available options
Color:
Blue
Blue, White
Camouflage
Gray, Black, White
Lime
Red, White
Red, White, Blue
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819507 Gray, Black, White
19819487 Red, White, Blue
19819488 Camouflage
19819489 Red, White
19819490 Blue, White
19819491 Blue
19819495 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19819507 Supplier Ergodyne™ Supplier No. 12359

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Evaporative Cooling Triangle Hat features activated polymers that provide cooling power in a variety of applications.

  • Activate by soaking in water for 2-5 minutes
  • Reusable
  • Tie closure
  • Available in a variety of colors and patterns like stars and stripes, camouflage, red western, navy western, blue, lime, flames, and skulls

Specifications

Product Type Cooling Hat
Color Gray, Black, White
Closure Type Tie
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling