missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ Chill-Its™ 6705 Evaporative Cooling Bandana with Hook and Loop
Description
- Soak in water 2-5 minutes to activate
- Remains hydrated for up to 4 hours
- Reusable
- Available in a variety of colors and patterns like red western, blue western, camo, stars and striped, black, lime, blue, skulls, and flames
Spécifications
Spécifications
| Product Type | Bandana |
| Color | Lime |
| Closure Type | Hook and Loop |
| For Use With (Application) | Construction, Maintenance, Landscaping, Material Handling |