missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6670CT Evap. Hard Hat Neck Shade with Cooling Towel

Catalog No. 19819544
Change view
Click to view available options
Color:
Lime
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819544 Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819544 Supplier Ergodyne™ Supplier No. 12523

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6670CT Evap. Hard Hat Neck Shade with Cooling Towel is designed to keep the sun off while delivering cooling neck comfort.

  • Advanced PVA material activates easily and quickly when run under water for one minute
  • Reusable
  • Evaporative cooling process helps keep you cool
  • Remains hydrated for up to 4 hours
  • Hook and loop straps for easy attachment

Specifications

Color Lime
For Use With (Application) Construction, Maintenance, Landscaping, Warehousing/Distribution
Material PVA