missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6660 Hard Hat Brim with Shade

Catalog No. 19819602
Change view
Click to view available options
Color:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819602 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819602 Supplier Ergodyne™ Supplier No. 12641

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hard Hat Brim with Shade fits around the outside of a hard hat to provide sun and UV protection on the face and neck.

  • 100% Polyester fabric
  • Universal design
  • Elastic inner rim to fit most hard hats
  • Designed to block sun and keep workers comfortable in outdoor environments

Specifications

Color Orange
For Use With (Application) Construction, Service, Landscaping, Material Handling
Material Polyester