missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6630 High-Performance Cap

Catalog No. 19154631
Change view
Click to view available options
Color:
Hi-vis Lime
Red Western
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19154631 Red Western
19154628 Hi-vis Lime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19154631 Supplier Ergodyne™ Supplier No. 12508

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Comfortable alone or under helmet

  • Hi Cool™ Moisture Management performance fabric
  • Comfortable to wear under hats, helmets, or alone
  • Wide, absorbent terrycloth band
  • Elastic band for a great fit

Specifications

Material Fabric
Product Type High Performance Cap
Color Red Western
For Use With (Application) Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication, hells kitchen
For Use With (Equipment) For use under hats, helmets, or alone
Includes Elastic Band