missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6630 High-Performance Cap

Catalog No. 19819545
Change view
Click to view available options
Color:
Black
Gray, Black, White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819545 Gray, Black, White
19819539 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19819545 Supplier Ergodyne™ Supplier No. 12529

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-Performance Cap constructed of technical fabric with a wide, absorbent terry headband is comfortable to wear under a helmet or alone.

  • Hi Cool™ moisture management performance fabric
  • Comfortable to wear under hats, helmets or alone
  • Wide, absorbent terrycloth band
  • One size fits most
  • Available in a varisty of colors and patterns like blue, black, skulls, and navy western

Specifications

Material Hi Cool™ Moisture Management Performance Fabric
Size Universal
Color Gray, Black, White
For Use With (Application) Construction, Maintenance, Landscaping, Distribution