missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6550 Head Sweatband

Catalog No. 19819521
Change view
Click to view available options
Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819521 Black
19819518 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19819521 Supplier Ergodyne™ Supplier No. 12452

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6550 Head Sweatband is constructed of thick soft terry to keep perspiration away from eyes and face.

  • Thick, absorbent cotton terry
  • Elastic maintains snug fit

Specifications

Color Black
For Use With (Application) Construction, Maintenance, Landscaping, Material Handling