missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6486 FR Multi-Band

Catalog No. 19819633
Change view
Click to view available options
Includes:
Anti-stink Treatment Helps Workers Remain Unscented
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19819633 Anti-stink Treatment Helps Workers Remain Unscented
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19819633 Supplier Ergodyne™ Supplier No. 42201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6486 FR Multi-Band provides FR protection and superior performance in a variety of applications.

  • Inherently FR Nomex™ knit fabric (EBT rating of 4.2 cal/cm2)
  • Meets requirements of Performance Specification ASTM F1506; classified CAT 1 according to NFPA 70E
  • Multiple style and usage configurations
  • Hi-tech moisture management performance fabric dries fast
  • Anti-stink treatment helps workers remain unscented
  • Multi-climate usage
  • Blocks wind, sun, and dust
  • Stretchable, comfortable, and seam-free
  • Machine washable
  • One size fits most

Specifications

Material Nomex Knit Fabric
Color Navy
Certifications/Compliance ASTM F1506, classified CAT 1 according to NFPA 70E
Includes Anti-stink Treatment Helps Workers Remain Unscented
For Use With (Application) Construction, Landscaping, Material Handling