missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6485 Multi-Band

Catalog No. 19819628
Change view
Click to view available options
Color:
Black, White
Blue, White
Hi-vis Orange
Hi-vis Yellow
Red, Black, White
White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19819628 Red, Black, White
19819617 Blue, White
19819618 Hi-vis Yellow
19819619 Hi-vis Orange
19819621 Black, White
19819607 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19819628 Supplier Ergodyne™ Supplier No. 42119

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chill-Its™ 6485 Multi-Band features supremely versatile performance fabric with a multitude of uses and ways to wear.

  • Multiple style and usage configurations
  • Hi-tech moisture management performance fabric dries fast
  • Multi-climate usage
  • Blocks wind, sun and dust
  • Stretchable, comfortable, and seam-free
  • Machine washable
  • Comes in the following patterns: Red Western, Navy Western, Hi-Vis Yellow, Hi-Vis Orange, Stars and Stripes, Flames, Skulls, Be Tenacious, Realtree™
  • One size

Specifications

Material Hi-tech Moisture Management Performance Fabric
Size Universal
Color Red, Black, White
For Use With (Application) Construction, Landscaping, Maintenance