missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ ChemSplash 1 Chemical Splash Sleeves with Taped Seams and Elastic Closures
Disposable, chemical splash sleeves are designed with red taped seams to protect your arms from chemical splashes and toxic particles.
Supplier: International Enviroguard™ 7065YT
Description
- Proprietary, hybrid membrane fabric helps keep arms cool as you work
- Secure, elastic closures on both ends help keep sleeves in place even with repetitive movements
- Longer length protects more of your arm from exposure to workplace hazards
- Taped seams offer an extra layer of protection by preventing contaminants from entering or seeping through exposed seams
- Elastic closures on both ends of sleeves keep them in place to prevent skin exposure
- High moisture vapor transfer rate prevents the buildup of heat and moisture to keep wearers cooler
- Possible applications include manufacturing, emergency response, environmental cleanup, and more
Specifications
| Chemical-resistant Sleeve | |
| Non-woven Fabric | |
| Non-sterile | |
| Single-use | |
| Yes | |
| 45.7 cm |
| Universal | |
| Yellow | |
| Elastic at Both Ends | |
| Stitched, Over-taped | |
| 18 in. |