missing translation for 'onlineSavingsMsg'
Learn More

Cesium, plasma standard solution, Specpure™ Cs 1000μg/mL

CAS: 534-17-8 | CCs2O3 | 325.82 g/mol

$98.71 - $322.08

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13825AD
View Documents
Thermo Scientific Chemicals
013825AD
50 mL
Each for $98.71
Only null left
 
AA13825AP
View Documents
Thermo Scientific Chemicals
013825AP
500 mL
Each for $322.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

534-17-8
325.82
FJDQFPXHSGXQBY-UHFFFAOYSA-L
[Cs+].[Cs+].[O-]C([O-])=O
CCs2O3
MFCD00010957
10796

Specifications

534-17-8 , 7697-37-2 , 7732-18-5
Liquid
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
dicesium;carbonate
50 mL
Nitric acid: 5.0%; Cesium carbonate: 0.122%; Water: 94.878%
23 hPa (17mm Hg) at 20°C
CCs2O3
Colorless
[Cs+].[Cs+].[O-]C([O-])=O
325.82
10796
Cesium
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only