accessibility menu, dialog, popup

UserName

CsCsOOO

CAS RN 534-17-8

IUPAC Name: dicaesium(1+) carbonate
Synonyms: caesium carbonate|carbonic acid, dicesium salt|cesium carbonate
Molecular Weight (g/mol): 325.82
Molecular Formula: CCs2O3
InChi Key: FJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES: [Cs+].[Cs+].[O-]C([O-])=O
Physical Form 
  • (7)
  • (2)
  • (11)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
Grade 
  • (5)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (2)
  • (20)
  • (7)
  • (2)
  • (11)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)