missing translation for 'onlineSavingsMsg'
Learn More

Cerium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLCE23X

 View more versions of this product

Catalog No. PLCE2-3X


Only null left

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ce10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Cerium Standard
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
MFCD00010924
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
GWXLDORMOJMVQZ-UHFFFAOYSA-N
5% HNO3
140.12
23974
Assurance
Triple-leached LDPE bottle
7440-45-1 , 7697-37-2 , 7732-18-5
Ce
AA, ICP
Certified Reference Material
[Ce]
cerium
500 mL
CHEBI:33369

Chemical Identifiers

7440-45-1
140.12
GWXLDORMOJMVQZ-UHFFFAOYSA-N
CHEBI:33369
[Ce]
Ce
MFCD00010924
23974
cerium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only