missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$417.72 - $887.00
Chemical Identifiers
| CAS | 7440-45-1 |
|---|---|
| Molecular Formula | Ce |
| Molecular Weight (g/mol) | 140.12 |
| MDL Number | MFCD00010924 |
| InChI Key | GWXLDORMOJMVQZ-UHFFFAOYSA-N |
| PubChem CID | 23974 |
| ChEBI | CHEBI:33369 |
| IUPAC Name | cerium |
| SMILES | [Ce] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLCE2-3Y
|
SPEX CertiPrep Group LLC
PLCE23Y |
125 mL | Triple-leached LDPE bottle |
Each for $417.72
|
|
||||
|
PLCE2-3X
|
SPEX CertiPrep Group LLC
PLCE23X |
500 mL | Triple-leached LDPE bottle |
Each for $887.00
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ce | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-45-1 | |
| 140.12 | |
| GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
| CHEBI:33369 | |
| [Ce] |
| Ce | |
| MFCD00010924 | |
| 23974 | |
| cerium |
Specifications
| Triple-leached LDPE bottle | |
| 7440-45-1 , 7697-37-2 , 7732-18-5 | |
| Ce | |
| AA, ICP | |
| Certified Reference Material | |
| [Ce] | |
| cerium | |
| 125 mL | |
| CHEBI:33369 | |
| Cerium Standard |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00010924 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
| 5% HNO3 | |
| 140.12 | |
| 23974 | |
| Assurance |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only