missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Oxycodone-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Numéro de catalogue. 01001802
Modifier l'affichage
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01001802 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01001802 Fournisseur Merck Emd Millipore Code fournisseur O0051ML
Il en reste null

Certified reference material

A stable-labeled internal standard suited for quantitation of oxycodone levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Oxycodone is a semi-synthetic opioid analgesic sold under many trade names including OxyContin™ and Percocet™ for treatment of moderate to severe pain. Oxycodone is also sold illicitly under street names such as 'Roxies,' 'Blues,' and 'Perks'.

Spécifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Linear Formula C18H18NO4D3
Concentration 100 μg/mL in methanol
MDL Number MFCD28038719
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 318.38

CERILLIANT is a registered trademark of Cerilliant Corporation