missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Flunitrazepam Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
A Certified Spiking Solution™ suitable for use in GC/MS or LC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. Flunitrazepam is an intermediate-acting benzodiazepine marketed under the trade name Rohypnol as a hypnotic, sedative, anticonvulsant and skeletal muscle relaxant.
Spécifications
Spécifications
| CAS | 67-56-1, 1622-62-4 |
| Linear Formula | C16H12FN3O3 |
| MDL Number | MFCD00057906 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 1.0 mg/mL in methanol |
| Formula Weight | 313.28 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation