missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Creatinine Solution, 2.0 mg/mL in Methanol:Water (1:1), Sold by MilliporeSigma™ Supelco™
Description
Creatinine is excreted at fairly constant rates in the absence of renal disease and commonly used as benchmark in urinary biomarker analysis. Serum creatinine levels are useful indicators of renal function and have been implicated in diabetes, cardiovascular and circulatory diseases. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls in creatine LC/MS or GC/MS testing methods for clinical diagnostic, urine drug testing or forensic analysis applications.
Spécifications
Spécifications
| CAS | 67-56-1, 60-27-5 |
| Linear Formula | C4H7N3O |
| MDL Number | MFCD00003203 |
| UN Number | UN 1992 6.1(3) / PGIII |
| Concentration | 2.0 mg/mL in methanol:water (1:1) |
| Formula Weight | 113.12 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 24°C (75.2°F) |
CERILLIANT is a registered trademark of Cerilliant Corporation