missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD CHO AGT™ Medium

Numéro de catalogue. p-7075674
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
12490017 1 L
12490025 10 L
12490001 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 12490017 Fournisseur Gibco™ Code fournisseur 12490017
Il en reste null

CD CHO AGT™ Medium

CD CHO AGT™ (Advanced Granulation Technology) medium is a protein-free, chemically-defined medium optimized for the growth of Chinese hamster ovary (CHO) cells and expression of recombinant proteins in suspension culture. The AGT™ dry media format is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. CD CHO AGT™ medium contains no proteins or peptide components of animal, plant, or synthetic origin, as well as no undefined lysates or hydrolysates.

When to Use CD CHO Medium
  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

    When to Consider Another Gibco™ Medium

    Consider CD FortiCHO™ medium when:
  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

    Consider CD OptiCHO™ medium when:
  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is finicky, i.e. hard to grow or adapt

    Consider CD DG44 medium when:
  • You use parental DG44 or DXB11 CHO cells (dhfr-)

    Liquid Media Formulations
    Gibco™ AGT™ media are also available in a liquid format that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.

    Design Your Own Media Specific to Your Needs
    Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
  • Spécifications

    Cell Line CHO
    Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Form Powder (AGT)
    Product Type CD CHO Medium
    Serum Level Serum-free
    With Additives Sodium Bicarbonate
    Without Additives No Glutamine, No Phenol Red, No Sodium Pyruvate, No Thymidine, No Hypoxanthine
    Product Line AGT, Gibco
    Quantity 1 L
    Shipping Condition Wet Ice
    Endotoxin Level ≤10 EU/mL, Low
    Format Securitainer Capped Vial
    Included Antibiotics No antibiotics
    Reconstituted Volume 1 L
    Species Hamster
    Content And Storage Store in refrigerator (2–8°C).
    Culture Type Suspension Cell Culture
    Afficher plus de résultats Afficher moins de résultats

    For Research Use Only. Not for use in diagnostic procedures.