missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Cassette II Biopsy Tissue Cassettes in Tube Packs

Numéro de catalogue. B851730OR
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Color
B851730OR Orange
B851730BE Beige
B851730BL Blue
B851730GN Green
B851730GY Gray
B851730LA Lavender
B851730LG Light Green
B851730LI Lilac
B851730TN Tan
B851730WH White
B851730YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Numéro de catalogue. B851730OR Fournisseur Epredia™ Code fournisseur B851730OR
Il en reste null

Biopsy Cassettes are ideal for small tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Spécifications

Pore Style Small Squares
Quantity 750/Cs.
Closure Included Yes
Closure Type Separate Lid, Front Hinge
PrintMate Compatible Yes
Color Orange
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Cassette II Biopsy Cassette