missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ LambdaXP Ligand Leakage ELISA
GREENER_CHOICE

Numéro de catalogue. 810375201
Change view
Click to view available options
Quantity:
1 Assay
10 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
810375201 1 Assay
810375210 10 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 810375201 Fournisseur Thermo Scientific™ Code fournisseur 810375201
Il en reste null

Thermo Scientific™ CaptureSelect™ LambdaXP Ligand Leakage ELISA detects possible presence of LambdaXP affinity ligand in the purified product after using CaptureSelect LambdaXP Affinity Matrix.

CaptureSelect LambdaXP Ligand Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using CaptureSelect LambdaXP Affinity Matrix.

The LambdaXP affinity ligand is a 13-kD recombinant protein, expressed in yeast and covalently coupled to a chromatography matrix to give high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach from the chromatography support and co-elute with the protein. This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect LambdaXP Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Spécifications

Capacity (Metric) 3 x 0.1 mL
Content And Storage -20°C
Shipping Condition Dry ice
Form Reagent Solution
Quantity 1 Assay
For Use With (Application) Affinity Ligand Leakage Quantitation Kits
Target CaptureSelect LambdaXP Ligand
Type ELISA Kit
Sufficient For 1 x 96-well plate