missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAT Conjugate
GREENER_CHOICE

Numéro de catalogue. 7102872500
Modifier l'affichage
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
7102872500 500 μg
7102872100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 7102872500 Fournisseur Thermo Scientific™ Code fournisseur 7102872500
Il en reste null

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAT Conjugate consists of a 13 kDa recombinant single domain antibody fragment that specifically binds with high affinity and selectivity to human alpha-1 antitrypsin (AAT).

CaptureSelect Biotin Anti-AAT Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human alpha-1 antitrypsin (AAT). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize native and recombinant human AAT.

Applications for CaptureSelect Biotin Anti-AAT Conjugate include ELISA, immunoprecipitation with streptavidin-coupled DynaBeads, Gyrolab™-based immunoassays, and label-free detection using platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Spécifications

Content And Storage Store in freezer (-5°C to -30°C).
Label Type Biotin and Analogs
Product Line CaptureSelect
Target Species Human
Ligand Type Antibody
Quantity 500 μg