missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ CapSeal Bullet™ Capillary Ferrule, General Purpose, 0.53 mm Column I.D.

Capillary ferrules have a precision bore which is centrally located. During assembly of a connector, fiber is fixed within the ferrule using adhesive before grinding and polishing for end preparation.

Supplier:  MilliporeSigma™ Supelco™ 23487

 Afficher plus de versions de ce produit

Numéro de catalogue. 111009096


Il en reste null

Description

Description

Spécifications

Identifiants chimiques

0.8 mm
Ferrule Type: General Purpose

Spécifications

0.8 mm
48/Pk.
Ferrule Type: General Purpose
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

CapSeal Bullet is a registered trademark of Sigma-Aldrich Co. LLC