missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ CapSeal Bullet™ Capillary Ferrule, General Purpose, 0.53 mm Column I.D.
Capillary ferrules have a precision bore which is centrally located. During assembly of a connector, fiber is fixed within the ferrule using adhesive before grinding and polishing for end preparation.
$202.63 - $533.88
Specifications
| Diameter (Metric) Inner | 0.8 mm |
|---|---|
| Description | Ferrule Type: General Purpose |
Specifications
| 0.8 mm | |
| Ferrule Type: General Purpose |
CapSeal Bullet is a registered trademark of Sigma-Aldrich Co. LLC