missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Burdick & Jackson™ Capping Reagent, 90/10 THF/NMI (v/v)
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Spécifications
Spécifications
| CAS | 109-99-9 |
| Content And Storage | Room Temperature |
| Density | 0.888 g/cm3 |
| Packaging Type | Glass Bottle |
| Quantity | 4 x 2 L |
| Form | Liquid |