missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ cAMP-Screen™ Wash Buffer

Catalog No. T2337
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2337 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2337 Supplier Invitrogen™ Supplier No. T2337

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with the cAMP-Screen or cAMP-Screen Direct™ Immunoassay system.

This Wash Buffer is suitable for use with the cAMP-Screen™ or cAMP-Screen Direct™ Immunoassay system.

  • Quantity: 1 L

Specifications

Product Type Wash Buffer
Quantity 1 L
Form Liquid
For Use With (Equipment) cAmp-Screen Direct™ Immunoassay System., cAMP-Screen™ Immunoassay System
Product Line cAMP-Screen
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.