Learn More
Parr™ Calorimeter Standards Benzoic Acid in Pellets
Calorific standard in pellet form for standardizing oxygen bomb calorimeters
Supplier:  Parr™ PAR3413
 Afficher plus de versions de ce produit
Numéro de catalogue. 04462
Il en reste null
Description
Description
Calorific standard in pellet form for standardizing oxygen bomb calorimeters
  • Traceable to NIST
Spécifications
Identifiants chimiques
Benzoic Acid
NIST
Standardizing the oxygen bomb calorimeters
Parr Oxygen Bomb Calorimeter
Spécifications
Benzoic Acid
Standardizing the oxygen bomb calorimeters
20/Bottle
1 g
NIST
Parr Oxygen Bomb Calorimeter
0.002 lb.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS