missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLCA22X

 View more versions of this product

Catalog No. PLCA2-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Calcium Standard
7440-70-2 , 7697-37-2 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
1.0,5.0,94.0
MFCD00010897 MFCD00085314
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
OYPRJOBELJOOCE-UHFFFAOYSA-N
2% HNO3
40.08
5460341
Assurance
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
1.0,5.0,94.0
Ca
AA, ICP
Certified Reference Material
[Ca]
calcium
500 mL
CHEBI:29320
Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLCA2-2Y, -2T, -2X
    10,000    PLCA2-3Y,-3X
2-5% HCl    1000μg/mL    PLCA1-2X
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only