missing translation for 'onlineSavingsMsg'
Learn More

Calcium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

Catalog No. p-5533047
Change view
Click to view available options
Quantity:
125 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PLCA22Y 125 mL
PLCA22T 250 mL
PLCA22X 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PLCA22Y Supplier SPEX CertiPrep Group LLC Supplier No. PLCA22Y
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Calcium Standard
Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS 7440-70-2, 7697-37-2, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 1.0, 5.0, 94.0
CAS Max % 1.0, 5.0, 94.0
Molecular Formula Ca
Technique AA, ICP
MDL Number MFCD00010897 MFCD00085314
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES [Ca]
Solvent or Matrix 2% HNO3
IUPAC Name calcium
Molecular Weight (g/mol) 40.08
Quantity 125 mL
PubChem CID 5460341
ChEBI CHEBI:29320
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLCA2-2Y, -2T, -2X
    10,000    PLCA2-3Y,-3X
2-5% HCl    1000μg/mL    PLCA1-2X
Show More Show Less

Product intended for laboratory use only