missing translation for 'onlineSavingsMsg'
Learn More

Calcium peroxide, typically 65%

Numéro de catalogue. p-7049753
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA2115722 100 g
AA2115736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA2115722 Fournisseur Thermo Scientific Chemicals Code fournisseur 02115722
Il en reste null

Rubber stabilizer | CAS: 1305-79-9 | CaO2 | 72.076 g/mol

Calcium peroxide is used as a rubber stabilizer, used in different areas of industry and agriculture. In agriculture it is used as an oxygen fertilizer, and is also used in the presowing treatments of rice seed. It is used as flour bleaching agent and improving agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 1305-79-9
Molecular Formula CaO2
Molecular Weight (g/mol) 72.076
MDL Number MFCD00015988
InChI Key LHJQIRIGXXHNLA-UHFFFAOYSA-N
Synonym calcium peroxide, calcium dioxide, calper, calper g, unii-7fro2eno91, peroxidocalcium, calciumperoxid, kalziumperoxid, calcium peroxide ca o2, hsdb 965
PubChem CID 14779
ChEBI CHEBI:48233
IUPAC Name calcium;peroxide
SMILES [O-][O-].[Ca+2]

Spécifications

Physical Form Powder
Quantity 100 g
UN Number UN1457
Merck Index 14,1689
Solubility Information Slightly soluble in water; Soluble in acids
Formula Weight 72.08
Percent Purity 65% (typically)
Odor Odorless
Sensitivity Moisture sensitive
Chemical Name or Material Calcium peroxide
Hazard Category H272-H314-H335
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CALCIUM PEROXIDE
EINECSNumber 215-139-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only