missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate tetrahydrate, MilliporeSigma™

Numéro de catalogue. p-5547623
Change view
Click to view available options
Quantity:
500 g
50 kg
Packaging:
Fibre carton
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
M1021210500 500 g Plastic Bottle
M1021219050 50 kg Fibre carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M1021210500 Fournisseur Technidata Code fournisseur 1.02121.0500
Il en reste null

For analysis EMSURE™ ACS

Identifiants chimiques

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Spécifications

Quantity 500 g
UN Number UN1454
Solubility Information Soluble in water: 2710g/L at 40°C
Formula Weight 236.15
Odor Odorless
Packaging Plastic Bottle
Density 1.82g/cm3 at 20°C
Chemical Name or Material Calcium nitrate tetrahydrate
Hazard Category H272: May intensify fire; oxidiser.
Hazard Statement H319: Causes serious eye irritation.
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-332-1
RTECSNumber EW3000000
Recommended Storage Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Away from combustible substances