missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Genistein, Soybean
A cell-permeable, reversible, substrate competitive inhibitor of protein tyrosine kinases, including autophosphorylation of epidermal growth factor receptor kinase (IC50 = 2.6 μM).
$199.63 - $199.63
Specifications
| Color | White |
|---|---|
| Target Proteins | EGFR kinase |
| CAS | 446-72-0 |
| Molecular Formula | C15H10O5 |
| Formula Weight | 270.2g/mol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
34-583-420MG
|
MilliporeSigma™
34583420MG |
20 mg |
Each for $199.63
|
|
|||||
Description
Cell PermeableSpecifications
| White | |
| 446-72-0 | |
| 270.2g/mol | |
| Protect from light | |
| 95% by HPLC |
| EGFR kinase | |
| C15H10O5 | |
| DMSO | |
| Solid |