missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Highly Purified Protease-Free RNase A, Bovine Pancreas

Catalog No. 55674650KU
Change view
Click to view available options
Quantity:
10 kU
50 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55674650KU 50 kU
55674610KU 10 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55674650KU Supplier MilliporeSigma™ Supplier No. 55674650KU

For selective degradation of RNA

  • Chromatographically purified
  • Pyrimidine-specific endoribonuclease that acts on single-stranded RNA

Specifications

Content And Storage -20°C
Source Bovine pancreas
Quantity 50 kU
Product Type Protease-Free RNase A
Form Lyophilized Solid

For Research Use Only