missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ High Purity IPTG, Dioxane-Free

Used in stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function

$121.00 - $5320.00

Specifications

Content And Storage 2- 8°C
Form White solid
Product Type IPTG
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
4203221GM
View Documents
MilliporeSigma™
4203221GM
1 g
Each for $121.00
Only null left
 
4203225GM
View Documents
MilliporeSigma™
4203225GM
5 g
Each for $323.00
Only null left
 
42032210GM
View Documents
MilliporeSigma™
42032210GM
10 g
Each for $440.00
Only null left
 
42032225GM
View Documents
MilliporeSigma™
42032225GM
25 g
Each for $900.00
Only null left
 
420322100GM
View Documents
MilliporeSigma™
420322100GM
100 g
Each for $2,200.00
Only null left
 
420322500GM
View Documents
MilliporeSigma™
420322500GM
500 g
Each for $5,320.00
Only null left
 
Description

Description

  • Contains ≤0.1% dioxane by GC
Specifications

Specifications

2- 8°C
White solid
IPTG
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only