missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ High Purity IPTG, Dioxane-Free

Used in stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function

Supplier:  MilliporeSigma™ 42032225GM

 View more versions of this product

Catalog No. 42032225GM


Only null left
Explore available promotions

Description

Description

  • Contains ≤0.1% dioxane by GC
Specifications

Specifications

2- 8°C
White solid
IPTG
25 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only