missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ High Purity IPTG, Dioxane-Free
Used in stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function
Supplier: MilliporeSigma™ 4203225GM
Description
- Contains ≤0.1% dioxane by GC
Specifications
| 2- 8°C | |
| White solid | |
| IPTG | |
| 5 g |
For Research Use Only