missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ FICOLL™ 400 Reagent, Molecular Biology Grade
Separation of lymphocyte subpopulations
$305.00 - $1,530.00
Specifications
| CAS | 26873-85-8 |
|---|---|
| Color | White |
| Content And Storage | Store at +15°C to +30°C |
| Formula Weight | 400000g/mol |
| For Use With (Application) | Separation of lymphocyte subpopulations |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
341691250GM
|
MilliporeSigma™
341691250GM |
250 g |
Each for $1,530.00
|
|
|||||
|
34169125GM
|
MilliporeSigma™
34169125GM |
25 g |
Each for $305.00
|
|
|||||
Description
Used for the separation of lymphocyte subpopulationsSpecifications
| 26873-85-8 | |
| Store at +15°C to +30°C | |
| Separation of lymphocyte subpopulations |
| White | |
| 400000g/mol | |
| Solid |