missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ FICOLL™ 400 Reagent, Molecular Biology Grade
Description
Used for the separation of lymphocyte subpopulations
Specifications
Specifications
| CAS | 26873-85-8 |
| Color | White |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 400,000 |
| Physical Form | Solid |
| For Use With (Application) | Separation of lymphocyte subpopulations |