missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ FICOLL™ 400 Reagent, Molecular Biology Grade

Catalog No. 341691250GM
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
341691250GM 250 g
34169125GM 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 341691250GM Supplier MilliporeSigma™ Supplier No. 341691250GM

Separation of lymphocyte subpopulations

Used for the separation of lymphocyte subpopulations

Specifications

CAS 26873-85-8
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 400000g/mol
For Use With (Application) Separation of lymphocyte subpopulations
Quantity 250 g
Form Solid