missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ DNase I, Bovine Pancreas

Acts on single and double stranded DNA

$366.00 - $709.00

Specifications

CAS 9003-98-9
Content And Storage Store at −20°C
Formula Weight 40000g/mol
Form Lyophilized
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
26091310MU
View Documents Promotion Details
MilliporeSigma™
26091310MU
10 mU
Each for $366.00
Only null left
 
26091325MU
View Documents Promotion Details
MilliporeSigma™
26091325MU
25 mU
Each for $709.00
Only null left
 
Description

Description

  • Native DNase I from bovine pancreas
  • An endonuclease that cleaves phosphodiester linkages adjacent to pyrimidine nucleotides to yield 5′-phosphate terminated polynucleotides with a free hydroxyl group at the 3′ position
Specifications

Specifications

9003-98-9
Store at −20°C
40000g/mol
Lyophilized
Videos
SDS
Documents

Documents