missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ DNase I, Bovine Pancreas

Catalog No. 26091310MU
Change view
Click to view available options
Quantity:
10 mU
25 mU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26091310MU 10 mU
26091325MU 25 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26091310MU Supplier MilliporeSigma™ Supplier No. 26091310MU

Acts on single and double stranded DNA

  • Native DNase I from bovine pancreas
  • An endonuclease that cleaves phosphodiester linkages adjacent to pyrimidine nucleotides to yield 5′-phosphate terminated polynucleotides with a free hydroxyl group at the 3′ position

Specifications

CAS 9003-98-9
Content And Storage Store at −20°C
Formula Weight 40000g/mol
MDL Number MFCD00130918
SMILES *
IUPAC Name Deoxyribonuclease
Molecular Weight (g/mol) 0.00
Quantity 10 mU
Form Lyophilized