missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ D(-)Mannitol

For Life Science research application.

$85.80 - $217.00

Specifications

CAS 69-65-8
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 182.2g/mol
Molecular Formula C6H14O6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
443907100GM
View Documents Promotion Details
MilliporeSigma™
443907100GM
100 g
Each for $85.80
Only null left
 
443907500GM
View Documents Promotion Details
MilliporeSigma™
443907500GM
500 g
Each for $217.00
Only null left
 
Description

Description

D(-)Mannitol - CAS 69-65-8 - Calbiochem
Specifications

Specifications

69-65-8
Store at +15°C to +30°C
C6H14O6
White
182.2g/mol
Crystalline Solid
Videos
SDS
Documents

Documents