missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ D(-)Mannitol

Catalog No. 443907100GM
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
443907100GM 100 g
443907500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. 443907100GM missing translation for 'mfr' MilliporeSigma™ missing translation for 'supplierNo' 443907100GM

For Life Science research application.

D(-)Mannitol - CAS 69-65-8 - Calbiochem

Specifications

CAS 69-65-8
Color White
Content And Storage Store at +15°C to +30°C
Formula Weight 182.2g/mol
Molecular Formula C6H14O6
Quantity 100 g
Form Crystalline Solid