missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Leupeptin, Hemisulfate, Microbial
Leupeptin hemisulfate, CAS 103476-89-7, is a reversible inhibitor of trypsin-like proteases and cysteine proteases.
Supplier: MilliporeSigma™ 10897550MG
Description
Peptide Sequence: Ac-Leu-Leu-arginal, hemisulfateSpecifications
| White | |
| 103476-89-7 | |
| 475.6g/mol | |
| Hygroscopic | |
| 50 mg |
| trypsin- and cysteine-like proteases | |
| C20H38N6O4·0.5H2SO4 | |
| H2O | |
| Lyophilized | |
| 90% by HPLC |