missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Leupeptin, Hemisulfate, Microbial

Catalog No. p-7060016
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1089755MG 5 mg
10897525MG 25 mg
10897550MG 50 mg
108975100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1089755MG Supplier MilliporeSigma™ Supplier No. 1089755MG
Only null left

Leupeptin hemisulfate, CAS 103476-89-7, is a reversible inhibitor of trypsin-like proteases and cysteine proteases.

Peptide Sequence: Ac-Leu-Leu-arginal, hemisulfate

Specifications

Color White
Target Proteins trypsin- and cysteine-like proteases
CAS 103476-89-7
Molecular Formula C20H38N6O4·0.5H2SO4
Formula Weight 475.6g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 5 mg
Purity 90% by HPLC